Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057876.5 | NP_477224.1 | 2 | 892 | 62 | |
NM_134311.3 | NP_599138.3 | 2 | 1009 | 62 | ||
GenBank mRNA | BT009968.1 | - | 2 | 883 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |