Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: disco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078638.4 | NP_523362.2 | 2 | 3170 | 140 | |
GenBank mRNA | BT003295.1 | - | 2 | 3155 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |