Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079178.3 | NP_523902.2 | 2 | 2275 | 69 | |
NM_168031.2 | NP_728881.1 | 2 | 1628 | 69 | ||
GenBank mRNA | BT150269.1 | - | 2 | 1629 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |