Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: viaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079308.2 | NP_524032.2 | 2 | 1039 | 136 | |
GenBank mRNA | AY122105.1 | - | 2 | 1015 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |