Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edg78E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079474.3 | NP_524198.1 | 2 | 879 | 143 | |
GenBank mRNA | M71248.1 | - | 2 | 833 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |