Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retinin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080256.4 | NP_524995.2 | 2 | 745 | 93 | |
GenBank mRNA | AY059445.1 | - | 919 | 93 | ||
BT025838.1 | - | 2629 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |