Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TotC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080518.3 | NP_536779.2 | 2 | 160 | 76 | |
GenBank mRNA | BT023305.1 | - | 2 | 143 | 76 | |
KX531366.1 | - | 2 | 116 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |