Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032110.2 | NP_001027281.1 | 3 | 1297 | 115 | |
NM_057561.6 | NP_476909.2 | 2 | 1178 | 115 | ||
NM_134291.5 | NP_599118.2 | 3 | 1374 | 115 | ||
NM_134292.3 | NP_599119.2 | 2 | 1159 | 115 | ||
NM_134293.3 | NP_599120.2 | 3 | 1355 | 115 | ||
GenBank mRNA | BT044503.1 | - | 2 | 1188 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |