Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ihog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135241.2 | NP_609085.1 | 2 | 3085 | 157 | |
GenBank mRNA | AY119465.1 | - | 2 | 3055 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |