Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299783.1 | NP_001286712.1 | 2 | 2799 | 139 | |
NM_137817.3 | NP_611661.1 | 3 | 3030 | 139 | ||
NM_166524.2 | NP_726187.1 | 3 | 2809 | 139 | ||
NM_166525.2 | NP_726188.1 | 3 | 3011 | 139 | ||
NM_166526.2 | NP_726189.1 | 2 | 2981 | 139 | ||
GenBank mRNA | AF395331.1 | - | 3 | 3007 | 139 | |
BT150349.1 | - | 3 | 3028 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |