Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccp84Ag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141420.3 | NP_649677.1 | 2 | 820 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |