Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccp84Aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141426.2 | NP_649683.1 | 2 | 632 | 135 | |
GenBank mRNA | BT011017.1 | - | 2 | 633 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |