Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF4E6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202377.1 | NP_001189306.1 | 2 | 617 | 127 | |
NM_001202378.1 | NP_001189307.1 | 2 | 607 | 127 | ||
NM_143397.2 | NP_651654.1 | 2 | 608 | 127 | ||
GenBank mRNA | AY094939.1 | - | 2 | 609 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster Inventoried |