Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143742.3 | NP_651999.1 | 3 | 1457 | 90 | |
GenBank mRNA | BT004476.1 | - | 3 | 1440 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |