Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kermit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299258.1 | NP_001286187.1 | 1 | 385 | 73 | |
NM_143771.4 | NP_652028.1 | 2 | 802 | 73 | ||
NM_165590.3 | NP_724649.1 | 2 | 443 | 73 | ||
NM_165591.3 | NP_724650.1 | 2 | 446 | 73 | ||
NM_165592.3 | NP_724651.1 | 2 | 330 | 73 | ||
GenBank mRNA | AY060451.1 | - | 2 | 723 | 73 | |
AY095011.1 | - | 2 | 277 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |