Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258821.2 | NP_001245750.1 | 3 | 1321 | 52 | |
NM_001258822.2 | NP_001245751.1 | 2 | 1457 | 52 | ||
NM_001258823.2 | NP_001245752.1 | 2 | 1643 | 52 | ||
NM_001298511.1 | NP_001285440.1 | 2 | 1291 | 52 | ||
NM_001298512.1 | NP_001285441.1 | 2 | 1633 | 52 | ||
NM_058103.3 | NP_477451.1 | 2 | 1636 | 52 | ||
NM_058104.5 | NP_477452.3 | 2 | 1636 | 52 | ||
NM_167651.2 | NP_728249.1 | 2 | 1241 | 52 | ||
NM_167652.2 | NP_728250.1 | 2 | 1243 | 52 | ||
NM_167653.3 | NP_728251.1 | 2 | 1453 | 52 | ||
NM_167654.2 | NP_728252.1 | 2 | 1172 | 52 | ||
NM_167655.2 | NP_728253.1 | 2 | 1240 | 52 | ||
NM_167656.2 | NP_728254.1 | 2 | 1238 | 52 | ||
GenBank mRNA | AB023511.1 | - | 2 | 1636 | 52 | |
AY122115.1 | - | 2 | 1638 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |