Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079225.3 | NP_523949.2 | 3 | 1230 | 65 | |
NM_168181.2 | NP_729191.1 | 2 | 1035 | 65 | ||
GenBank mRNA | AF367369.1 | - | 3 | 1216 | 65 | |
AY095010.1 | - | 3 | 1209 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |