Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274987.1 | NP_001261916.1 | 2 | 689 | 57 | |
NM_001274988.1 | NP_001261917.1 | 3 | 1239 | 57 | ||
NM_001274989.1 | NP_001261918.1 | 2 | 1168 | 57 | ||
NM_079377.3 | NP_524101.2 | 2 | 953 | 57 | ||
NM_168644.2 | NP_730097.1 | 2 | 805 | 57 | ||
NM_168645.2 | NP_730098.1 | 2 | 755 | 57 | ||
GenBank mRNA | BT031197.1 | - | 2 | 787 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |