Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104173.3 | NP_001097643.1 | 3 | 1278 | 86 | |
NM_001144499.3 | NP_001137971.1 | 7 | 1427 | 86 | ||
NM_001275107.1 | NP_001262036.1 | 6 | 1175 | 86 | ||
NM_001275108.2 | NP_001262037.1 | 3 | 1278 | 86 | ||
NM_001275109.1 | NP_001262038.1 | 3 | 603 | 86 | ||
NM_079431.2 | NP_524155.1 | 2 | 503 | 86 | ||
NM_168787.3 | NP_730389.1 | 3 | 669 | 86 | ||
GenBank mRNA | BT023785.1 | - | 3 | 1256 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |