Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141777.4 | NP_650034.1 | 3 | 1063 | 75 | |
NM_169372.3 | NP_731527.1 | 2 | 1162 | 75 | ||
GenBank mRNA | AY069712.1 | - | 2 | 647 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |