Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ravus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169498.3 | NP_731779.1 | 2 | 1157 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |