Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sur-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275735.1 | NP_001262664.1 | 3 | 1188 | 60 | |
NM_001275736.1 | NP_001262665.1 | 3 | 1416 | 60 | ||
NM_142363.7 | NP_650620.3 | 3 | 1188 | 60 | ||
NM_169758.4 | NP_732231.2 | 3 | 1416 | 60 | ||
GenBank mRNA | AY128499.1 | - | 3 | 1413 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |