Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp53C14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014532.3 | NP_001014532.1 | 2 - 3 | 206 | 63 | |
NM_001202032.2 | NP_001188961.1 | 2 - 3 | 153 | 63 | ||
GenBank mRNA | BT032710.1 | - | 2 - 3 | 180 | 63 | |
KX532105.1 | - | 1 - 2 | 44 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |