Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014589.4 | NP_001014589.1 | 4 - 5 | 822 | 60 | |
GenBank mRNA | BT031129.1 | - | 4 - 5 | 788 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |