Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sinu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139714.4 | NP_647971.3 | 1 - 2 | 853 | 110 | |
GenBank mRNA | AY423544.1 | - | 1 - 2 | 816 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |