Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipped-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042986.3 | NP_001036451.1 | 22 - 23 | 6342 | 76 | |
NM_001042987.3 | NP_001036452.1 | 22 - 23 | 6370 | 76 | ||
NM_001169581.2 | NP_001163052.1 | 23 - 24 | 6263 | 76 | ||
NM_001169582.2 | NP_001163053.1 | 23 - 24 | 6298 | 76 | ||
NM_001169583.2 | NP_001163054.1 | 23 - 24 | 6270 | 76 | ||
NM_001273774.1 | NP_001260703.1 | 21 - 22 | 6010 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |