Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038805.2 | NP_001033894.1 | 2 - 3 | 430 | 74 | |
GenBank mRNA | AY226541.1 | - | 2 - 3 | 235 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |