Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038805.2 | NP_001033894.1 | 5 - 6 | 982 | 126 | |
GenBank mRNA | AY226541.1 | - | 5 - 6 | 787 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |