Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038805.2 | NP_001033894.1 | 9 - 10 | 1423 | 89 | |
GenBank mRNA | AY226541.1 | - | 9 - 10 | 1228 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |