Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104161.3 | NP_001097631.1 | 7 - 8 | 1434 | 76 | |
NM_001104162.4 | NP_001097632.1 | 2 - 3 | 955 | 76 | ||
NM_001144489.3 | NP_001137961.1 | 2 - 3 | 955 | 76 | ||
NM_001259883.2 | NP_001246812.1 | 2 - 3 | 955 | 76 | ||
NM_001259884.2 | NP_001246813.1 | 2 - 3 | 955 | 76 | ||
NM_001275050.2 | NP_001261979.1 | 7 - 8 | 1434 | 76 | ||
NM_168718.3 | NP_730245.2 | 7 - 8 | 1434 | 76 | ||
GenBank mRNA | AY122226.1 | - | 7 - 8 | 1432 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |