Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbp80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015151.3 | NP_001015151.1 | 9 - 10 | 1181 | 96 | |
NM_001276239.1 | NP_001263168.1 | 9 - 10 | 1181 | 96 | ||
NM_001276240.1 | NP_001263169.1 | 8 - 9 | 1134 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |