Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015157.4 | NP_001015157.2 | 7 - 8 | 1020 | 56 | |
NM_001015158.4 | NP_001015158.1 | 6 - 7 | 957 | 56 | ||
NM_001015159.4 | NP_001015159.2 | 3 - 4 | 2320 | 56 | ||
NM_001015160.4 | NP_001015160.2 | 7 - 8 | 984 | 56 | ||
NM_001015161.4 | NP_001015161.2 | 5 - 6 | 933 | 56 | ||
NM_001015162.4 | NP_001015162.1 | 7 - 8 | 884 | 56 | ||
NM_001110995.2 | NP_001104465.1 | 6 - 7 | 1195 | 56 | ||
NM_001201439.2 | NP_001188368.1 | 8 - 9 | 1132 | 56 | ||
NM_001201440.1 | NP_001188369.1 | 7 - 8 | 1222 | 56 | ||
GenBank mRNA | BT082094.1 | - | 3 - 4 | 2281 | 56 | |
BT120108.1 | - | 7 - 8 | 841 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |