Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111011.3 | NP_001104481.2 | 1 - 2 | 281 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |