Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015319.2 | NP_001015319.1 | 4 - 5 | 1387 | 89 | |
NM_001276232.1 | NP_001263161.1 | 3 - 4 | 1326 | 89 | ||
GenBank mRNA | BT015266.1 | - | 4 - 5 | 1350 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |