Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015389.3 | NP_001015389.1 | 3 - 4 | 297 | 115 | |
NM_001276231.1 | NP_001263160.1 | 3 - 4 | 338 | 115 | ||
GenBank mRNA | AY118788.1 | - | 2 - 3 | 275 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |