Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015498.5 | NP_001015498.5 | 7 - 8 | 2416 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |