Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042973.3 | NP_001036438.1 | 14 - 15 | 2090 | 97 | |
NM_001169580.2 | NP_001163051.1 | 15 - 16 | 2005 | 97 | ||
NM_001299206.1 | NP_001286135.1 | 14 - 15 | 2090 | 97 | ||
GenBank mRNA | BT022770.1 | - | 14 - 15 | 2023 | 97 | |
BT022801.1 | - | 14 - 15 | 2065 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |