Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crabp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001842.1 | NP_001001842.1 | 3 - 4 | 466 | 93 | |
GenBank mRNA | AY616861.1 | - | 3 - 4 | 466 | 93 | |
BC162180.1 | - | 3 - 4 | 443 | 93 | ||
BC162190.1 | - | 3 - 4 | 443 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |