Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 77 - 78 | 16608 | 79 | |
XM_005165511.4 | XP_005165568.1 | 77 - 78 | 16727 | 79 | ||
XM_009301753.3 | XP_009300028.1 | 78 - 79 | 16749 | 79 | ||
GenBank mRNA | AY540131.1 | - | 77 - 78 | 16612 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |