Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 79 - 80 | 17200 | 88 | |
XM_005165511.4 | XP_005165568.1 | 79 - 80 | 17319 | 88 | ||
XM_009301753.3 | XP_009300028.1 | 80 - 81 | 17341 | 88 | ||
GenBank mRNA | AY540131.1 | - | 79 - 80 | 17204 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |