Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003640.2 | NP_001003640.1 | 4 - 5 | 594 | 65 | |
GenBank mRNA | BC078304.1 | - | 4 - 5 | 598 | 65 | |
KC136847.1 | - | 3 - 4 | 387 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |