Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004494.1 | NP_001004494.1 | 13 - 14 | 1792 | 55 | |
XM_021470440.1 | XP_021326115.1 | 13 - 14 | 1869 | 55 | ||
GenBank mRNA | BC075789.1 | - | 13 - 14 | 1792 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |