Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007280.1 | NP_001007281.1 | 1 - 2 | 98 | 106 | |
XM_021467980.1 | XP_021323655.1 | 1 - 2 | 125 | 106 | ||
GenBank mRNA | AY351958.1 | - | 1 - 2 | 71 | 106 | |
BC085411.1 | - | 1 - 2 | 98 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |