Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smtnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013321.1 | NP_001013339.1 | 2 - 3 | 712 | 65 | |
XM_005157213.4 | XP_005157270.1 | 2 - 3 | 756 | 65 | ||
XM_021481009.1 | XP_021336684.1 | 2 - 3 | 757 | 65 | ||
GenBank mRNA | BC090695.1 | - | 2 - 3 | 712 | 65 | |
BC114242.1 | - | 2 - 3 | 686 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |