Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013456.1 | NP_001013474.1 | 4 - 5 | 261 | 102 | |
GenBank mRNA | BC091561.1 | - | 4 - 5 | 261 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |