Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013469.1 | NP_001013487.1 | 3 - 4 | 300 | 81 | |
GenBank mRNA | BC091463.1 | - | 3 - 4 | 300 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |