Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020537.1 | NP_001018373.1 | 11 - 12 | 1596 | 71 | |
XM_009305736.3 | XP_009304011.1 | 11 - 12 | 1619 | 71 | ||
XM_021479616.1 | XP_021335291.1 | 11 - 12 | 1628 | 71 | ||
XM_021479617.1 | XP_021335292.1 | 11 - 12 | 1606 | 71 | ||
GenBank mRNA | BC095090.1 | - | 11 - 12 | 1596 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |