Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020537.1 | NP_001018373.1 | 12 - 13 | 1666 | 96 | |
XM_009305736.3 | XP_009304011.1 | 12 - 13 | 1689 | 96 | ||
XM_021479616.1 | XP_021335291.1 | 12 - 13 | 1698 | 96 | ||
XM_021479617.1 | XP_021335292.1 | 12 - 13 | 1676 | 96 | ||
GenBank mRNA | BC095090.1 | - | 12 - 13 | 1666 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |