Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vtna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020672.1 | NP_001018508.1 | 3 - 4 | 542 | 86 | |
XM_021481411.1 | XP_021337086.1 | 4 - 5 | 86 | |||
GenBank mRNA | BC095724.1 | - | 3 - 4 | 542 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |