Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024397.1 | NP_001019568.1 | 13 - 14 | 1372 | 68 | |
GenBank mRNA | BC095257.1 | - | 13 - 14 | 1372 | 68 | |
BC165709.1 | - | 13 - 14 | 1387 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |